Merck
Simultaneous measurement of T
Magnetic resonance in medicine (2017-05-19)
Eric Aliotta, Kévin Moulin, Zhaohuan Zhang, Daniel B Ennis
ABSTRAKT
To evaluate a technique for simultaneous quantitative T T Phantom T T
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)