Merck
Molecular Engineering of Mn
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-11-23)
Chiara Maccato, Lorenzo Bigiani, Giorgio Carraro, Alberto Gasparotto, Roberta Seraglia, Jiyeon Kim, Anjana Devi, Gloria Tabacchi, Ettore Fois, Giuseppe Pace, Vito Di Noto, Davide Barreca
ABSTRAKT
Molecular engineering of manganese(II) diamine diketonate precursors is a key issue for their use in the vapor deposition of manganese oxide materials. Herein, two closely related β-diketonate diamine Mn