Merck
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-09-01)
Kwang Bo Jung, Hana Lee, Ye Seul Son, Ji Hye Lee, Hyun-Soo Cho, Mi-Ok Lee, Jung-Hwa Oh, Jaemin Lee, Seokho Kim, Cho-Rok Jung, Janghwan Kim, Mi-Young Son
ABSTRAKT
Human intestinal organoids (hIOs) derived from human pluripotent stem cells (hPSCs) have immense potential as a source of intestines. Therefore, an efficient system is needed for visualizing the stage of intestinal differentiation and further identifying hIOs derived from hPSCs. Here, 2 fluorescent biosensors were developed based on human induced pluripotent stem cell (hiPSC) lines that stably expressed fluorescent reporters driven by intestine-specific gene promoters Krüppel-like factor 5 monomeric Cherry (KLF5
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DMH1, ≥98% (HPLC)