Merck
  • Resistance Monitoring of Four Insecticides and a Description of an Artificial Diet Incorporation Method for Chilo suppressalis (Lepidoptera: Crambidae).
Resistance Monitoring of Four Insecticides and a Description of an Artificial Diet Incorporation Method for Chilo suppressalis (Lepidoptera: Crambidae).
Journal of economic entomology (2017-10-19)
Huang Shuijin, Chen Qiong, Qin Wenjing, Sun Yang, Qin Houguo
ABSTRAKT
Chilo suppressalis (Walker; Lepidoptera: Crambidae) is one of the most damaging rice pests in China. Insecticides play a major role in its management. We describe how we monitored the resistance of C. suppressalis to four insecticides in seven field populations from Jiangxi, Hubei, and Hunan Provinces, China, in 2014-2016. The topical application method for resistance monitoring was suitable for triazophos, monosultap, and abamectin. The conventional rice seedling dipping method proved ineffective for testing chlorantraniliprole so the new artificial diet incorporation method was substituted. This new method provided more consistent results than the other methods, once baseline toxicity data had been established. All populations had moderate to high resistance to triazophos from 2014 to 2016. Monosultap resistance in two populations increased from low in 2014 to moderate in 2016 and the other five populations showed moderate to high-level resistance throughout. Abamectin resistance in three populations increased from sensitive or low in 2014 to moderate in 2015-2016, and the other populations had moderate to high levels of resistance. Resistance to chlorantraniliprole increased from sensitive or low in 2014 to moderate to high in 2016. These results suggested that resistance management strategies should be developed according to the needs of a specific location. It was suggested that, in these localities, organophosphate insecticides should be prohibited, the application of nereistoxin, macrolide antibiotic, and diamide insecticides should be limited, and other insecticides, including spinetoram and methoxyfenozide, that exhibited no resistance should be used rationally and in rotation to delay resistance development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Chlorantraniliprole, PESTANAL®, analytical standard
Supelco
Flubendiamide, PESTANAL®, analytical standard