Merck
  • Pharmacological analysis of the transmembrane action potential configuration in myoepithelial cells of the spontaneously beating heart of the ascidian
Pharmacological analysis of the transmembrane action potential configuration in myoepithelial cells of the spontaneously beating heart of the ascidian
The Journal of experimental biology (2017-10-07)
Vladimir A Golovko, Igor A Kosevich, Mikhail A Gonotkov
ABSTRAKT
The mechanisms of action potential (AP) generation in the myoepithelial cells of the tunicate heart are not yet well understood. Here, an attempt was made to elucidate these mechanisms by analyzing the effects of specific blockers of K
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade