Merck
The Immunosuppressant Mycophenolic Acid Alters Nucleotide and Lipid Metabolism in an Intestinal Cell Model.
Scientific reports (2017-03-23)
Svenja Heischmann, Monika Dzieciatkowska, Kirk Hansen, Dieter Leibfritz, Uwe Christians
ABSTRAKT
The study objective was to elucidate the molecular mechanisms underlying the negative effects of mycophenolic acid (MPA) on human intestinal cells. Effects of MPA exposure and guanosine supplementation on nucleotide concentrations in LS180 cells were assessed using liquid chromatography-mass spectrometry. Proteomics analysis was carried out using stable isotope labeling by amino acids in cell culture combined with gel-based liquid chromatography-mass spectrometry and lipidome analysis using
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-ACSL5 antibody produced in mouse, clone 5H8, purified immunoglobulin, buffered aqueous solution