Merck
Casein kinase II phosphorylation of cyclin F at serine 621 regulates the Lys48-ubiquitylation E3 ligase activity of the SCF
Open biology (2017-10-13)
Albert Lee, Stephanie L Rayner, Alana De Luca, Serene S L Gwee, Marco Morsch, Vinod Sundaramoorthy, Hamideh Shahheydari, Audrey Ragagnin, Bingyang Shi, Shu Yang, Kelly L Williams, Emily K Don, Adam K Walker, Katharine Y Zhang, Justin J Yerbury, Nicholas J Cole, Julie D Atkin, Ian P Blair, Mark P Molloy, Roger S Chung
ABSTRAKT
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disorder that is characterized by progressive weakness, paralysis and muscle loss often resulting in patient death within 3-5 years of diagnosis. Recently, we identified disease-linked mutations in the
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone D66, purified from hybridoma cell culture