- Activation-Dependent TRAF3 Exon 8 Alternative Splicing Is Controlled by CELF2 and hnRNP C Binding to an Upstream Intronic Element.
Activation-Dependent TRAF3 Exon 8 Alternative Splicing Is Controlled by CELF2 and hnRNP C Binding to an Upstream Intronic Element.
Molecular and cellular biology (2016-12-30)
Astrid-Solveig Schultz, Marco Preussner, Mario Bunse, Rotem Karni, Florian Heyd
PMID28031331
ABSTRAKT
Cell-type-specific and inducible alternative splicing has a fundamental impact on regulating gene expression and cellular function in a variety of settings, including activation and differentiation. We have recently shown that activation-induced skipping of TRAF3 exon 8 activates noncanonical NF-κB signaling upon T cell stimulation, but the regulatory basis for this splicing event remains unknown. Here we identify
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CUGBP2 antibody, Mouse monoclonal, clone HL1889 (1H2-1F12), purified from hybridoma cell culture