Merck
The effects of gestational age and maternal hypoxia on the placental renin angiotensin system in the mouse.
Placenta (2014-09-25)
J S M Cuffe, S L Walton, S E Steane, R R Singh, D G Simmons, K M Moritz
ABSTRAKT
The renin angiotensin system (RAS) is an important mediator of placental development. However, a comprehensive expression profile for 8 key components of the placental RAS throughout murine gestation has not been performed. Furthermore, maternal hypoxia induces dysregulation of RAS expression in fetal tissues but the effects on the murine placental RAS are less well known. Placentas were collected from male and female CD1 mouse fetuses at seven gestational ages for qPCR analysis of Agt, Ren1, Atp6ap2, Ace, Ace2, Agtr1a, Agtr2 and Mas1. mRNA localisation of Agtr1 and Mas1 and protein localisation of ACE and ACE2 was determined at E18.5. To determine the effects of maternal hypoxia on the placental RAS, mice were housed in 12% oxygen from E14.5-E18.5 and placentas examined at E18.5. All RAS genes were expressed in the placenta throughout pregnancy and expression varied with fetal sex and age. Agtr1 was expressed within the labyrinth while Mas1 was expressed within the intraplacental yolk sac. ACE and ACE2 were localised to both labyrinth and junctional zones. In response to maternal hypoxia the expression of Agt, Ace and Ace2 was decreased but expression of Agtr1a was increased. Ace and Agtr1a mRNA levels were affected to a greater extent in females compared to males. Collectively, the location within the placenta as well as the expression profiles identified, support a role for the placental RAS in labyrinth development. The placental RAS is disturbed by maternal hypoxia in a sexually dimorphic manner and may contribute to impairment of placental vascular development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture