Merck
Local influence of high molecular polyethylene particles on heterotopic ossification.
Experimental and therapeutic medicine (2017-06-08)
Lin-Ru Zeng, Fang-Bing Zhu, Jian-Yue Wang, Qiao Hou, Zhen-Shuang Yue, Shi-Gui Yan, Ren-Fu Quan, Ying-Liang Zhang
ABSTRAKT
We studied the effect of molecular polyethylene particles on local heterotopic ossification. A total of 36 healthy Sprague-Dawley rats were randomly divided into the control group (n=18) and the observation group (n=18). High molecular polyethylene particles were injected to rupture Achilles tendon position in the observation group, and normal saline was injected in the control group. X-ray examinations were conducted on Achilles tendon in the 4th, 8th and 12th week after operation. The incidence rate of heterotopic ossification was evaluated, and bone trabecula morphological structure was studied under optical microscope after hematoxylin and eosin staining. Bone morphogenetic protein 2 (BMP-2), transforming growth factor-β (TGF-β), interleukin-1 (IL-1), tumor necrosis factor-α (TNF-α), runt-related transcription factor 2 (Runx2) and matrix metalloproteinase-9 (MMP-9) expression levels were also measured. Our results showed that heterotopic ossification incidence in the observation group was significantly lower than that in the control group. Achilles tendon structure in the control group increased in volume, and its texture was harder and cartilage-like. In the observation group, trabecular bone volume, thickness and quantity were more than those observed in the control group. BMP-2, TGF-β, IL-1, TNF-α, Runx2 and MMP-9 levels in the observation group were significantly lower than those in the control group. We concluded that, high molecular polyethylene particles had a significant inhibiting effect on local heterotopic ossification.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-CBFA1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MMP9 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution