Merck
Id genes are essential for early heart formation.
Genes & development (2017-08-11)
Thomas J Cunningham, Michael S Yu, Wesley L McKeithan, Sean Spiering, Florent Carrette, Chun-Teng Huang, Paul J Bushway, Matthew Tierney, Sonia Albini, Mauro Giacca, Miguel Mano, Pier Lorenzo Puri, Alessandra Sacco, Pilar Ruiz-Lozano, Jean-Francois Riou, Muriel Umbhauer, Gregg Duester, Mark Mercola, Alexandre R Colas
ABSTRAKT
Deciphering the fundamental mechanisms controlling cardiac specification is critical for our understanding of how heart formation is initiated during embryonic development and for applying stem cell biology to regenerative medicine and disease modeling. Using systematic and unbiased functional screening approaches, we discovered that the Id family of helix-loop-helix proteins is both necessary and sufficient to direct cardiac mesoderm formation in frog embryos and human embryonic stem cells. Mechanistically, Id proteins specify cardiac cell fate by repressing two inhibitors of cardiogenic mesoderm formation-Tcf3 and Foxa2-and activating inducers Evx1, Grrp1, and Mesp1. Most importantly, CRISPR/Cas9-mediated ablation of the entire Id (Id1-4) family in mouse embryos leads to failure of anterior cardiac progenitor specification and the development of heartless embryos. Thus, Id proteins play a central and evolutionarily conserved role during heart formation and provide a novel means to efficiently produce cardiovascular progenitors for regenerative medicine and drug discovery applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid