Merck
Spatiotemporal patterning of EpCAM is important for murine embryonic endo- and mesodermal differentiation.
Scientific reports (2018-01-31)
Sannia Sarrach, Yuanchi Huang, Sebastian Niedermeyer, Matthias Hachmeister, Laura Fischer, Sebastian Gille, Min Pan, Brigitte Mack, Gisela Kranz, Darko Libl, Juliane Merl-Pham, Stefanie M Hauck, Elisa Paoluzzi Tomada, Matthias Kieslinger, Irmela Jeremias, Antonio Scialdone, Olivier Gires
ABSTRAKT
Epithelial cell adhesion molecule EpCAM is expressed in pluripotent embryonic stem cells (ESC) in vitro, but is repressed in differentiated cells, except epithelia and carcinomas. Molecular functions of EpCAM, possibly imposing such repression, were primarily studied in malignant cells and might not apply to non-pathologic differentiation. Here, we comprehensively describe timing and rationale for EpCAM regulation in early murine gastrulation and ESC differentiation using single cell RNA-sequencing datasets, in vivo and in vitro models including CRISPR-Cas9-engineered ESC-mutants. We demonstrate expression of EpCAM in inner cell mass, epiblast, primitive/visceral endoderm, and strict repression in the most primitive, nascent Flk1+ mesoderm progenitors at E7.0. Selective expression of EpCAM was confirmed at mid-gestation and perinatal stages. The rationale for strict patterning was studied in ESC differentiation. Gain/loss-of-function demonstrated supportive functions of EpCAM in achieving full pluripotency and guided endodermal differentiation, but repressive functions in mesodermal differentiation as exemplified with cardiomyocyte formation. We further identified embryonic Ras (ERas) as novel EpCAM interactor of EpCAM and an EpCAM/ERas/AKT axis that is instrumental in differentiation regulation. Hence, spatiotemporal patterning of EpCAM at the onset of gastrulation, resulting in early segregation of interdependent EpCAM+ endodermal and EpCAM-/vimentin+ mesodermal clusters represents a novel regulatory feature during ESC differentiation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys27) - przeciwciało i zestaw primerów zweryfikowane pod kątem ChIP, from rabbit, purified by using Protein A
Sigma-Aldrich
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys4) - przeciwciało z walidacją ChIP i zestaw primerów, królicze monoklonalne, from rabbit