Merck
  • Effect of different mini-volume colloid centrifugation configurations on flow cytometrically sorted sperm recovery efficiency and quality using a computer-assisted semen analyzer.
Effect of different mini-volume colloid centrifugation configurations on flow cytometrically sorted sperm recovery efficiency and quality using a computer-assisted semen analyzer.
Reproduction in domestic animals = Zuchthygiene (2017-09-12)
L B Ferré, J L Chitwood, C Fresno, H H Ortega, M E Kjelland, P J Ross
ABSTRAKT
Straws of sex-sorted sperm are usually packaged at a low concentration (e.g., ~2.1 × 10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hypotaurine, ≥98% (TLC)