Merck
Comparison of a new nanoform of the photosensitizer chlorin e6, based on plant phospholipids, with its free form.
FEBS open bio (2018-02-13)
Lyubov V Kostryukova, Vladimir N Prozorovskiy, Natalya V Medvedeva, Olga M Ipatova
ABSTRAKT
Photodynamic therapy is an advanced method of treating cancer and various benign diseases, including infections. It uses light-activated molecules [photosensitizers (PSs)] to generate reactive oxygen species (ROS) when irradiated with light of a specific wavelength. This study examined the photophysical and photosensitizing activity of the PS chlorin e6 incorporated in a delivery system based on plant phospholipids. This new nanoform of chlorin e6 comprised particles with a diameter of 18.4 ± 2.5 nm and zeta potential of -34.6 ± 3.0 mV. Incorporation of chlorin e6 in phospholipid nanoparticles was observed to cause a bathochromic shift of Q-band absorption maximum by 14 nm without an absorption change in the range of the Soret band. Fluorescence intensity of chlorin e6 embedded in the phospholipid nanoparticles increased 1.7-fold. Chlorin e6 in phospholipid nanoparticles, when irradiated, was able to generate ROS as shown by oxidation of polyunsaturated fatty acids of the phospholipid matrix of the delivery system and reduced l-glutathione.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology