Merck
Association of diabetes mellitus and biochemical knee cartilage composition assessed by T
Journal of magnetic resonance imaging : JMRI (2017-05-31)
Nattagan Chanchek, Alexandra S Gersing, Benedikt J Schwaiger, Michael C Nevitt, Jan Neumann, Gabby B Joseph, Nancy E Lane, Julia Zarnowski, Felix C Hofmann, Ursula Heilmeier, Charles E McCulloch, Thomas M Link
ABSTRAKT
To investigate the association of the presence and severity of diabetes mellitus (DM) with articular cartilage composition, using magnetic resonance imaging (MRI)-based T In the Osteoarthritis Initiative 208, participants with DM (age 63.0 ± 8.9 years; 111 females) and risk factors for osteoarthritis (OA) or mild radiographic tibiofemoral OA (Kellgren-Lawrence [KL] grade ≤2) were identified and group-matched with 208 controls without DM (age 63.3 ± 9.1 years; 111 females). Subjects with diabetes-related renal or ophthalmological complications or insulin treatment at baseline (n = 50) were defined as severe DM. 3T MR images of the right knee were assessed for articular cartilage T DM subjects had significantly higher cartilage T In comparison to nondiabetic controls, cartilage in DM subjects showed higher and more heterogeneous cartilage T 2 Technical Efficacy: 5 J. Magn. Reson. Imaging 2018;47:380-390.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)