- Roles of hydroxyl and sulfate radicals in degradation of trichloroethene by persulfate activated with Fe
Roles of hydroxyl and sulfate radicals in degradation of trichloroethene by persulfate activated with Fe
Journal of hazardous materials (2017-10-17)
Yunde Liu, Aiguo Zhou, Yiqun Gan, Xiaoqian Li
PMID29032099
ABSTRAKT
Active species including hydroxyl (HO) and sulfate radicals (SO