Merck
CDKN2B deletion is essential for pancreatic cancer development instead of unmeaningful co-deletion due to juxtaposition to CDKN2A.
Oncogene (2017-09-12)
Q Tu, J Hao, X Zhou, L Yan, H Dai, B Sun, D Yang, S An, L Lv, B Jiao, C Chen, R Lai, P Shi, X Zhao
ABSTRAKT
Pancreatic cancer is among the deadliest malignancies; however, the genetic events that lead to pancreatic carcinogenesis in adults remain unclear. In vivo models in which these genetic alterations occur in adult animals may more accurately reflect the features of human cancer. In this study, we demonstrate that inactivation of Cdkn2b (p15ink4b) is necessary for induction of pancreatic cancer by oncogenic KRAS
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Polyethylenimine, branched, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched