Merck
Loss of p53-inducible long non-coding RNA
Oncotarget (2017-12-22)
Markus Kaller, Ursula Götz, Heiko Hermeking
ABSTRAKT
We have previously identified the long non-coding RNA
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution