- Nanoencapsulated Illicium verum Hook.f. essential oil as an effective novel plant-based preservative against aflatoxin B
Nanoencapsulated Illicium verum Hook.f. essential oil as an effective novel plant-based preservative against aflatoxin B
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2017-11-12)
Abhishek Kumar Dwivedy, Vipin Kumar Singh, Bhanu Prakash, Nawal Kishore Dubey
PMID29126800
ABSTRAKT
The study reports efficacy of Illicium verum essential oil (IvEO) against food borne moudls and its nanoencapsulation for enhancing antifungal and antiaflatoxigenic potency. Chemical characterization of the IvEO showed anethole (89.12%) as major compound followed by estragole (4.859%). The IvEO showed broad fungitoxic spectrum against common food borne moulds. It's minimum inhibitory concentration (MIC) and minimum aflatoxin B