- Long non-coding RNA PTENP1 interacts with miR-193a-3p to suppress cell migration and invasion through the PTEN pathway in hepatocellular carcinoma.
Long non-coding RNA PTENP1 interacts with miR-193a-3p to suppress cell migration and invasion through the PTEN pathway in hepatocellular carcinoma.
Oncotarget (2018-01-04)
Yu-Yuan Qian, Kun Li, Quan-Yan Liu, Zhi-Su Liu
PMID29296207
ABSTRAKT
Long non-coding RNA PTENP1, the pseudogene of PTEN tumor suppressor, was previously reported to be a tumour suppressor in some cancer types. However, the precise effects mediated by PTENP1 transcripts within intricate regulatory networks involving molecular interactions with PTEN and tumorigenicity in hepatocellular carcinoma (HCC) remains elusive. Here, we identify the critical biological functions of PTENP1 and discuss whether PTENP1 could directly interact with miR-193a-3p to affect the progression of HCC both
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA
Informacje o cenach i dostępności nie są obecnie dostępne.