Merck
Polyphenols isolated from Acacia mearnsii bark with anti-inflammatory and carbolytic enzyme inhibitory activities.
Chinese journal of natural medicines (2018-01-14)
Jia Xiong, Mary H Grace, Debora Esposito, Slavko Komarnytsky, Fei Wang, Mary Ann Lila
ABSTRAKT
The present study was designed to characterize the polyphenols isolated from Acacia mearnsii bark crude extract (B) and fractions (B1-B7) obtained by high-speed counter-current chromatography (HSCCC) and evaluate their anti-inflammatory and carbolytic enzymes (α-glucosidase and α-amylase) inhibitory activities. Fractions B4, B5, B6, B7 (total phenolics 850.3, 983.0, 843.9, and 572.5 mg·g
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Supelco
Procyanidin B2, analytical standard