Merck
  • P2X receptor overexpression induced by soluble oligomers of amyloid beta peptide potentiates synaptic failure and neuronal dyshomeostasis in cellular models of Alzheimer's disease.
P2X receptor overexpression induced by soluble oligomers of amyloid beta peptide potentiates synaptic failure and neuronal dyshomeostasis in cellular models of Alzheimer's disease.
Neuropharmacology (2017-10-29)
Francisco Sáez-Orellana, María C Fuentes-Fuentes, Pamela A Godoy, Tiare Silva-Grecchi, Jessica D Panes, Leonardo Guzmán, Gonzalo E Yévenes, Javiera Gavilán, Terrance M Egan, Luis G Aguayo, Jorge Fuentealba
ABSTRAKT
The most common cause of dementia is Alzheimer's disease. The etiology of the disease is unknown, although considerable evidence suggests a critical role for the soluble oligomers of amyloid beta peptide (Aβ). Because Aβ increases the expression of purinergic receptors (P2XRs) in vitro and in vivo, we studied the functional correlation between long-term exposure to Aβ and the ability of P2XRs to modulate network synaptic tone. We used electrophysiological recordings and Ca
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
8-Cyclopentyl-1,3-dipropylxanthine, solid