Merck
A novel filtration system for point of care washing of cellular therapy products.
Journal of tissue engineering and regenerative medicine (2016-10-07)
Rui Tostoes, John R Dodgson, Ben Weil, Spyridon Gerontas, Chris Mason, Farlan Veraitch
ABSTRAKT
The cell therapy industry would greatly benefit from a simple point of care solution to remove dimethylsulphoxide (DMSO) from small-volume thawed cell suspensions before injection. A novel dead-end filtration device has been designed and validated, which takes advantage of the higher density of thawed cell suspensions to remove the DMSO and protein impurities from the cell suspension without fouling the filter membrane. The filter was designed to avoid fluid circuits and minimize the surface area that is contacted by the cell suspension, thus reducing cell losses by design. The filtration process was established through optimization of the fluid flow configuration, backflush cycles and filter geometry. Overall, this novel filtration device allows for a 1 ml of thawed cryopreserved cell suspensions, containing 10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Aminobenzoyl-Ala-Phe-Ala-Phe-Asp-Val-Phe-Tyr(NO2)-Asp, >90% (HPLC)