Merck
Validation of a simultaneous method for determining polycyclic aromatic compounds and alkylated isomers in biota.
Rapid communications in mass spectrometry : RCM (2017-11-28)
Ifeoluwa Idowu, Olga Francisco, Philippe J Thomas, Wesley Johnson, Chris Marvin, Jörg Stetefeld, Gregg T Tomy
ABSTRAKT
There is a need for a validated method to improve detection limits and simultaneously quantify polycyclic aromatic compounds (PACs, both parent and alkylated homologues) in biota by gas chromatography/tandem mass spectrometry because of their environmental significance. The validation of the method was performed in accordance to the Eurachem Guide to Quality in Analytical Chemistry. Gas chromatography coupled with a triple quadrupole mass spectrometer used in multiple reaction monitoring (MRM) mode was used for detection and quantification. Retention time windows and selective MRM ion transitions were optimized for a suite of PACs. The developed method was validated by comparing our measurements made on a reference material of freeze-dried mussel tissue (Mytilus edulis) with the certified values. Linearity was observed between 10-1000 pg/μL (PAHs) and 2-500 pg/μL (alkyl-PACs including S-based PACs). The overall mean (±SD) for the limits of detection of 43 PACs studied were 0.305 ± 0.276 and 2.69 ± 1.10 ng/g, respectively. For the 14 certified target analytes, the percent relative error ranged from 1.3 to 33%. With the exception of benzo(a)pyrene, the between-day and within-day repeatability for all target analytes was lower than 15% RSD. This is the first report of a fully validated method to simultaneously quantify PACs in biota performed in an ISO accredited laboratory.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1-Methylpyrene, suitable for fluorescence, ≥97.0% (GC)
Sigma-Aldrich
4-Methyldibenzothiophene, 96%