Merck
Polysaccharide lyases.
Applied biochemistry and biotechnology (1986-04-01)
R J Linhardt, P M Galliher, C L Cooney
ABSTRAKT
Polysaccharide lyases (or eliminases) are a class of enzymes (EC 4.2.2.-) that act to cleave certain activated glycosidic linkages present in acidic polysaccharides. These enzymes act through an eliminase mechanism, rather than through hydrolysis, resulting in unsaturated oligosaccharide products. Acidic polysaccharides are ubiquitous and so are the lyases that degrade them. This review article examines lyases that act on acidic polysaccharides of plant, animal, and microbial origin. These lyases are predominantly of microbial origin and come from a wide variety of both pathogenic and nonpathogenic bacteria and fungi. The lyases discussed include alginate lyase (EC 4.2.2.3), pectin lyase (EC 4.2.2.10), pectate lyase (EC 4.2.2.2), oligogalacturonide lyase (EC 4.2.2.6), exopolygalacturonate lyase (EC 4.2.2.9), chondroitin lyases (EC 4.2.2.4 and EC 4.2.2.5), hyaluronate lyase (EC 4.2.2.1), heparin lyase (EC 4.2.2.7), heparan lyase (EC 4.2.2.8), and other unclassified lyases. This review examines the sources, regulation, purification, and properties of these polysaccharide lyases.