Merck
Cannabinoids determination in oral fluid by SPME-GC/MS and UHPLC-MS/MS and its application on suspected drivers.
Science & justice : journal of the Forensic Science Society (2014-12-17)
Luca Anzillotti, Erika Castrignanò, Sabina Strano Rossi, Marcello Chiarotti
ABSTRAKT
The confirmation of Δ9-tetrahydrocannabinol (THC) in oral fluid (OF) is an important issue for assessing Driving Under the Influence of Drugs (DUID). The aim of this research was to develop a highly sensitive method with minimal sample pre-treatment suitable for the analysis of small OF volumes (100 μL) for the confirmation of cannabinoids in DUID cases. Two methods were compared for the confirmation of THC in residual OF samples, obtained from a preliminary on-site screening with commercial devices. An ultra high performance LC-MS (UHPLC-MS/MS) method and an SPME-GC/MS method were hence developed. 100 μL of the residual mixture OF/preservative buffer or neat OF was simply added to 10 μL of THC-D3 (1 μg/mL) and submitted to the two different analyses: A - direct injection of 10 μL in UHPLC-MS/MS in positive electrospray ionisation (ESI) mode and B - sampling for 30 min with SPME (100 μm polydimethylsiloxane or PDMS fibre) and direct injection by desorption of the fibre in the GC injection port. The lowest limit of detection (LLOD) of THC was 2 ng/mL in UHPLC-MS/MS and 0.5 ng/mL in SPME-GC/MS. In addition, cannabidiol (CBD) and cannabinol (CBN) could be detected in GC/MS equipment at 2 ng/mL, whilst in UHPLC-MS/MS the LLOD was 20 ng/mL. Both methods were applied to 70 samples coming from roadside tests. By SPME-GC/MS analysis, THC was confirmed in 42 samples, whilst CBD was detected in 21 of them, along with CBN in 14 samples. THC concentrations ranged from traces below the lowest limit of quantification or LLOQ (2 ng/mL) up to 690 ng/mL.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Overcoated Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, notched pink hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4