Merck
Passive monitors to measure hydrogen sulfide near concentrated animal feeding operations.
Environmental science. Processes & impacts (2013-05-18)
Brian T Pavilonis, Patrick T O'Shaughnessy, Ralph Altmaier, Nervana Metwali, Peter S Thorne
ABSTRAKT
Hydrogen sulfide (H2S) is one of many airborne pollutants emitted by concentrated animal feeding operations (CAFOs). However, few studies have characterized ambient H2S levels near these facilities, largely due to the lack of low-cost, reliable, and easily transportable instrumentation available to researchers. We determined intermediate environmental H2S exposure near CAFOs using Radiello passive monitors. First, a laboratory study was performed to determine the accuracy of the device. Next, a total of eight passive H2S monitors were deployed bi-weekly in close proximity (<40 m) to a medium-sized swine confinement for seven months in order to determine the temporal and spatial variability of H2S. Finally, we measured H2S concentrations across two rural Iowa counties to characterize ambient exposure near thirteen CAFOs and two schools. The value of the temperature-adjusted H2S passive diffusion rate provided by the supplier was 29% larger than the 24 h rate determined experimentally. Concentrations of H2S measured near the medium-sized confinement were varied and ranged from 0.2 to 48.6 ppb depending on the sampling period and proximity to a lagoon on the property. Two-week concentrations near the schools were low (<1 ppb), while concentrations near the thirteen CAFOs ranged from 0.1 to 42.9 ppb. The passive monitors were effective in measuring H2S concentrations near a swine CAFO as long as they were exposed for a sufficient period of time (two weeks). Radiello passive monitors are a promising new device in measuring intermediate H2S exposure in rural populations. Measured values in excess of an Iowa state limit of 30 ppb (24 h average) suggest that enforcement actions are needed to mitigate H2S migration from swine CAFOs.
MATERIAŁY
Numer produktu
Marka
Opis produktu
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
radiello Clips, suspending Radiello triangular support plate, pk of 20
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
radiello BTEX/VOC Starter Kit, CS2 Desorption
radiello BTEX/VOC Starter Kit, Thermal Desorption
radiello On-Field Thermometer, Reader, pk of 1
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
radiello Diffusive Bodies, white, configured for general use, pk of 20
Supelco
radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
Supelco
radiello BTEX Calibration Kit (Thermal Desorption)
radiello Filtration Kit, pk of 20
Supelco
radiello Aldehyde Calibration Standard, 50 μg/mL each component in acetonitrile, pkg of 10 mL
Supelco
radiello BTEX Calibration Kit (CS2 Desorption)
radiello Zestaw startowy siarkowodoru
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
radiello Triangular Support Plate, pk of 20
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
radiello Barcode Labels, pk of 198
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
radiello Vertical Adapter, threaded for standard use, pk of 20
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
radiello Outdoor Shelter, pk of 10
radiello Anesthetic Gases and Vapor Sampler
radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100