Merck
  • Field comparison of passive air samplers with reference monitors for ambient volatile organic compounds and nitrogen dioxide under week-long integrals.
Field comparison of passive air samplers with reference monitors for ambient volatile organic compounds and nitrogen dioxide under week-long integrals.
Journal of environmental monitoring : JEM (2009-01-13)
Shaibal Mukerjee, Karen D Oliver, Robert L Seila, Henry H Jacumin, Carry Croghan, E Hunter Daughtrey, Lucas M Neas, Luther A Smith
ABSTRAKT
This study evaluates performance of nitrogen dioxide (NO2) and volatile organic compound (VOC) passive samplers with corresponding reference monitors at two sites in the Detroit, Michigan area during the summer of 2005. Ogawa passive NO2 samplers and custom-made, re-useable Perkin-Elmer (PE) tubes with Carbopack X sorbent for VOCs were deployed under week-long sampling periods for six weeks. Precise results (5% relative standard deviation, RSD) were found for NO2 measurements from collocated Ogawa samplers. Reproducibility was also good for duplicate PE tubes for benzene, toluene, ethylbenzene, and xylene isomers (BTEX species, all < or = 6% RSD). As seen in previous studies, comparison of Ogawa NO2 samplers with reference chemiluminescence measurements suggested good agreement. Generally good agreement was also found between the PE tubes and reference methods for BTEX species.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Carbopack B, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack X, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack X, 40-60 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 60-80 mesh, bottle of 10 g
Supelco
Carbopack Adsorbent, matrix Carbopack C, 80-100 mesh, bottle of 10 g
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack B, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., unconditioned, pkg of 10 ea, matrix Carbopack
Supelco
Carbopack B, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
FLM Carbopack X Deactivated Stainless Steel TD Tube, preconditioned, pkg of 10 tubes, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
Supelco
Carbopack X, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbopack Adsorbent, matrix Carbopack B, 60-80 mesh, bottle of 10 g