Merck
Study of inhibitors of the PGH synthases for which potency is regulated by the redox state of the enzymes.
Methods in molecular biology (Clifton, N.J.) (2010-07-21)
Olivier Boutaud, John A Oates
ABSTRAKT
Aspirin, salicylic acid, and acetaminophen are examples of drugs that inhibit the PGH synthases (cyclooxygenases) with potencies that vary remarkably between different cells. This results from the fact that the inhibitory action of these drugs is regulated by redox cycling of the enzymes, which is determined by the concentration of hydroperoxides in the enzyme proximity. The potency of these drugs is greatest in the presence of low concentrations of hydroperoxides. Accordingly, analysis of the action of these inhibitors requires consideration of assay conditions that take into account the factors that control hydroperoxide concentrations, both in studies with purified enzymes and in cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
Interferon-γ from mouse, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture