- A new chemical tool for absinthe producers, quantification of α/β-thujone and the bitter components in Artemisia absinthium.
A new chemical tool for absinthe producers, quantification of α/β-thujone and the bitter components in Artemisia absinthium.
Food chemistry (2016-07-28)
Benoit Bach, Marilyn Cleroux, Mayra Saillen, Patrik Schönenberger, Stephane Burgos, Julien Ducruet, Armelle Vallat
PMID27451252
ABSTRAKT
The concentrations of α/β-thujone and the bitter components of Artemisia absinthium were quantified from alcoholic wormwood extracts during four phenological stages of their harvest period. A solid-phase micro-extraction method coupled to gas chromatography-mass spectrometry was used to determine the concentration of the two isomeric forms of thujone. In parallel, the combination of ultra-high pressure liquid chromatography and high resolution mass spectrometry allowed to quantify the compounds absinthin, artemisetin and dihydro-epi-deoxyarteannuin B. This present study aimed at helping absinthe producers to determine the best harvesting period.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub