Skip to Content
MilliporeSigma

Skip To

AV40323

Anti-CUGBP2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CUG triplet repeat, RNA binding protein 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL
Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
$522.00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

human, rat, horse, rabbit, mouse, bovine, dog, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CUGBP2(10659)

General description

CUGBP2 (ETR-3/NAPOR/BRUNOL3) is an RNA-binding protein, posttranscriptional controller of gene expression, that regulates key cellular responses such as apoptosis by binding to AU-rich sequences in the mRNA of important genes such as COX-2 and VEGF. CUGBP2 is a critical regulator of the apoptotic response to genotoxic injury in breast cancer cells and mitotic catastrophe response.

Immunogen

Synthetic peptide directed towards the N terminal region of human CUGBP2

Application

Anti-CUGBP2 polyclonal antibody is used to tag RNA binding protein 2 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of RNA binding protein 2 in the regulation of posttranslational gene transcription in response to gentotoxic injury.

Biochem/physiol Actions

Anti-CUGBP2 polyclonal antibody reacts with chicken, human, mouse, rat, canine, bovine, and zebrafish RNA binding protein 2 proteins.
Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation.Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Alternative splicing results in multiple transcript variants encoding different isoforms.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Other Notes

Synthetic peptide located within the following region: VYQINVLRDRSQNPPQSKGCCFVTFYTRKAALEAQNALHNIKTLPGMHHP

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV40324SAB1408786SAB2103977
description

IgG fraction of antiserum

description

affinity isolated antibody

description

purified immunoglobulin, buffered aqueous solution

description

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

Gene Information

human ... CUGBP2(10659)

Gene Information

human ... CUGBP2(10659)

Gene Information

human ... CUGBP1(10658)

Gene Information

human ... CUGBP1(10658)

mol wt

56 kDa

mol wt

56 kDa

mol wt

antigen 51.6 kDa

mol wt

56 kDa


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters