Skip to Content
Merck

04401

Ammonium hypophosphite

≥97.0%

Synonym(s):

Ammonium phosphinate

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
NH4H2PO2
CAS Number:
Molecular Weight:
83.03
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

≥97.0%

impurities

≤0.001% heavy metals (as Pb)

anion traces

chloride (Cl-): ≤100 mg/kg
phosphite (H3PO3): ≤1%
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤5 mg/kg
Ca: ≤500 mg/kg
Fe: ≤10 mg/kg

SMILES string

N.OPO

InChI

1S/H3N.H3O2P/c;1-3-2/h1H3;1-3H

InChI key

ZRDXIILBYDTYOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel synthesis of Ni2 P/MCM-41 catalysts by reducing a precursor of ammonium hypophosphite and nickel chloride at low temperature.
Song H, et al.
Applied Catalysis A: General, 462, 247-255 (2013)
Yung-Kuan Tseng et al.
Journal of nanoscience and nanotechnology, 14(10), 7907-7910 (2015-05-07)
In this study, the hydrothermal method is used to grow phosphorus-doped ZnO nanowires on Si/SiO2 substrates deposited with Al-doped ZnO thin film. This structure forms a homogeneous p-n junction. In this study, we are the pioneers to use ammonium hypophosphite
Zachariasen WH and Mooney RCL.
J. Chem. Phys. , 2(1), 34-37 (1934)
The ESR spectrum of irradiated ammonium hypophosphite.
Morton J R.
Molecular Physics, 5(3), 217-223 (1962)
L F Rozhko et al.
Amino acids, 29(2), 139-143 (2005-05-10)
The method for the synthesis of 2-substituted 2-hydroxycarbonylethyl-1-aminoalkylphosphinic acids (I) (pseudo-alpha,alpha-dipeptides) from ammonium and potassium hypophosphites (II) is described. The proposed route to the synthesis of pseudo-alpha,alpha-dipeptides consists in addition hypophosphite to acrylic compounds and formation of the first phosphorus-carbon

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service