Skip to Content

BDPH9BCD5B95

Bide Pharmatech

Ethyl 2-Fluoro-6-methoxybenzoate

Synonym(s):

ethyl 2-fluoro-6-methoxybenzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H11O3F1
CAS Number:
Molecular Weight:
198.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H11FO3/c1-3-14-10(12)9-7(11)5-4-6-8(9)13-2/h4-6H,3H2,1-2H3

Inchi Key

PWTGADHDRWQHRI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Ethyl 2-Fluoro-6-methoxybenzoate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library