Skip to Content
Merck

SAB1404138

Monoclonal Anti-NPY, (C-terminal) antibody produced in mouse

clone 2C10, purified immunoglobulin, buffered aqueous solution

Synonym(s):

PYY4

Sign In to View Organizational & Contract Pricing

Select a Size

100 μG

4 480,00 kr

4 480,00 kr


Please contact Customer Service for Availability


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~33.7 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPY(4852)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA044572AB9608SAB1404134
conjugate

unconjugated

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

species reactivity

human

species reactivity

human

species reactivity

rat, human

species reactivity

human

Gene Information

human ... NPY(4852)

Gene Information

human ... NPY(4852)

Gene Information

human ... NPY(4852)

Gene Information

human ... NME3(4832)

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

mouse

clone

2C10, monoclonal

clone

polyclonal

clone

polyclonal

clone

3G10, monoclonal

Immunogen

NPY (AAH29497, 29 a.a. ~ 97 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
YPSKPDNPGEDAPAEDMARYYSALRHYINLITRQRYGKRSSPETLISDLLMRESTENVPRTRLEDPAMW

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elysia M S Tan et al.
Acta paediatrica (Oslo, Norway : 1992), 106(2), 292-297 (2016-11-28)
We investigated the expression of neuropeptide Y (NPY), NPY receptor 1 (NPYR1) and NPY receptor 2 (NPYR2) in infantile haemangiomas (IHs). Immunohistochemical (IHC) staining was performed on proliferating IHs from six patients aged 4-13 (mean 8.7) months and involuted IHs
Bin Qiu et al.
Journal of genetics and genomics = Yi chuan xue bao, 43(7), 421-430 (2016-07-28)
Neuropeptide Y (NPY) is widely expressed in the central nervous system and influences many physiological processes. It is located within the rat quantitative trait locus (QTL) for alcohol preference on chromosome 4. Alcohol-nonpreferring (NP) rats consume very little alcohol, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service