Skip to Content
Merck

COM448679032

Combi-Blocks, Inc.

Methyl 3-(2,5,6-trimethoxypyridin-3-yl)-4,5-dihydroisoxazole-5-carboxylate

Sign In to View Organizational & Contract Pricing

Select a Size

Product COM448679032 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
296.28
MDL number:
Product COM448679032 is not currently sold in your country.

Iupac Name

methyl 3-(2,5,6-trimethoxy-3-pyridinyl)-2,5-dihydro-5-isoxazolecarboxylate

Inchi Code

1S/C13H16N2O6/c1-17-9-5-7(11(18-2)14-12(9)19-3)8-6-10(21-15-8)13(16)20-4/h5-6,10,15H,1-4H3

InChI key

AQEPLSBOVBRZCZ-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?ST-0880

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?ST-0880


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library