Skip to Content
Merck

749044

Titanium

sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
Ti
CAS Number:
Molecular Weight:
47.87
UNSPSC Code:
12352103
PubChem Substance ID:
EC Number:
231-142-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Titanium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.995% trace metals basis

InChI key

RTAQQCXQSZGOHL-UHFFFAOYSA-N

InChI

1S/Ti

SMILES string

[Ti]

assay

99.995% trace metals basis

form

solid

autoignition temp.

860 °F

reaction suitability

core: titanium

resistivity

42.0 μΩ-cm, 20°C

diam. × thickness

3.00 in. × 0.125 in.

bp

3287 °C (lit.)

mp

1660 °C (lit.)

density

4.5 g/mL at 25 °C (lit.)

Application

Sputtering is a process whereby atoms are ejected from a solid target material due to bombardment of the target by energetic particles. It is commonly used for thin-film deposition, etching and analytical techniques.

Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dahn, J. R.; Trussler, S.; Hatchard, T. D.; Bonakdarpour, B. et al.
Chemistry of Materials, 14, 3519-3519 (2002)
Hahn, H.; Averback, R. S.
Journal of Applied Physics, 67, 1113-1113 (1990)
Sigmund, P.
Physical Review, 184, 383-383 (1969)
Oechsner, H.
Applied Physics. A, Materials Science & Processing, 8, 185-185 (1975)
A Kurbad et al.
International journal of computerized dentistry, 16(2), 125-141 (2013-08-13)
This article presents two novel options for lithium-disilicate restorations supported by single-tooth implants. By using a Ti-Base connector, hybrid abutments and hybrid abutment crowns can be fabricated for different implant systems. The latter option in particular is an interesting new

Articles

คุณสมบัติของอุปกรณ์จำนวนมากถูกจำกัดโดยคุณสมบัติภายในของวัสดุที่ประกอบกัน

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Nanocomposite Coatings with Tunable Properties Prepared by Atomic Layer Deposition

Spintronics offer breakthroughs over conventional memory/logic devices with lower power, leakage, saturation, and complexity.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service