Skip to Content
Merck

BDPH9BCD080F

Bide Pharmatech

2,4-Difluorobenzoyl chloride

Synonym(s):

2,4-difluorobenzoyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1Cl1F2
CAS Number:
Molecular Weight:
176.55
MDL number:
Product BDPH9BCD080F is not currently sold in your country.


Inchi Code

1S/C7H3ClF2O/c8-7(11)5-2-1-4(9)3-6(5)10/h1-3H

Inchi Key

JSWRVDNTKPAJLB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Colorless to white to Yellow Liquid or Semi-Solid or solid



Still not finding the right product?

Explore all of our products under 2,4-Difluorobenzoyl chloride


hstatements

signalword

Danger

pictograms

CorrosionExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library