Skip to Content
Merck

CRM963

PCB Congeners - Silty Loam

pkg of 30 g

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

PCB Congeners - Silty Loam, pkg of 30 g

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 30 g

manufacturer/tradename

RTC CRM963-030

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service