Merck
  • Kinetics of 1,2-dichloroethane and 1,2-dibromoethane biodegradation in anaerobic enrichment cultures.
Kinetics of 1,2-dichloroethane and 1,2-dibromoethane biodegradation in anaerobic enrichment cultures.
Applied and environmental microbiology (2012-12-25)
Rong Yu, Hari S Peethambaram, Ronald W Falta, Matthew F Verce, James K Henderson, Christopher E Bagwell, Robin L Brigmon, David L Freedman
ABSTRACT
1,2-Dichloroethane (1,2-DCA) and 1,2-dibromoethane (ethylene dibromide [EDB]) contaminate groundwater at many hazardous waste sites. The objectives of this study were to measure yields, maximum specific growth rates (μ), and half-saturation coefficients (K(S)) in enrichment cultures that use 1,2-DCA and EDB as terminal electron acceptors and lactate as the electron donor and to evaluate if the presence of EDB has an effect on the kinetics of 1,2-DCA dehalogenation and vice versa. Biodegradation was evaluated at the high concentrations found at some industrial sites (>10 mg/liter) and at lower concentrations found at former leaded-gasoline sites (1.9 to 3.7 mg/liter). At higher concentrations, the Dehalococcoides yield was 1 order of magnitude higher when bacteria were grown with 1,2-DCA than when they were grown with EDB, while μ's were similar for the two compounds, ranging from 0.19 to 0.52 day(-1) with 1,2-DCA to 0.28 to 0.36 day(-1) for EDB. K(S) was larger for 1,2-DCA (15 to 25 mg/liter) than for EDB (1.8 to 3.7 mg/liter). In treatments that received both compounds, EDB was always consumed first and adversely impacted the kinetics of 1,2-DCA utilization. Furthermore, 1,2-DCA dechlorination was interrupted by the addition of EDB at a concentration 100 times lower than that of the remaining 1,2-DCA; use of 1,2-DCA did not resume until the EDB level decreased close to its maximum contaminant level (MCL). In lower-concentration experiments, the preferential consumption of EDB over 1,2-DCA was confirmed; both compounds were eventually dehalogenated to their respective MCLs (5 μg/liter for 1,2-DCA, 0.05 μg/liter for EDB). The enrichment culture grown with 1,2-DCA has the advantage of a more rapid transition to 1,2-DCA after EDB is consumed.
Product Description
Supelco
1,2-Dichloroethane, analytical standard
Supelco
1,2-Dibromoethane solution, certified reference material, 200 μg/mL in methanol
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dibromoethane, 98%