Merck
  • Activin A contributes to the development of hyperoxia-induced lung injury in neonatal mice.
Activin A contributes to the development of hyperoxia-induced lung injury in neonatal mice.
Pediatric research (2015-03-12)
Rebecca Lim, Ruth Muljadi, Eugenia Koulaeva, Patricia Vosdoganes, Siow Teng Chan, Rutu Acharya, Seshini Gurusinghe, Olli Ritvos, Arja Pasternack, Euan M Wallace
ABSTRACT
Bronchopulmonary dysplasia (BPD) is one of the leading causes of morbidity and mortality in babies born prematurely, yet there is no curative treatment. In recent years, a number of inhibitors against TGFβ signaling have been tested for their potential to prevent neonatal injury associated with hyperoxia, which is a contributing factor of BPD. In this study, we assessed the contribution of activin A-a member of the TGFβ superfamily-to the development of hyperoxia-induced lung injury in neonatal mice. We placed newborn C57Bl6 mouse pups in continuous hyperoxia (85% O2) to mimic many aspects of BPD including alveolar simplification and pulmonary inflammation. The pups were administered activin A receptor type IIB-Fc antagonist (ActRIIB-Fc) at 5 mg/kg or follistatin at 0.1 mg/kg on postnatal days 4, 7, 10, and 13. Treatment with ActRIIB-Fc and follistatin protected against hyperoxia-induced growth retardation. ActRIIB-Fc also reduced pulmonary leukocyte infiltration, normalized tissue: airspace ratio and increased septal crest density. These findings were associated with reduced phosphorylation of Smad3 and decreased matrix metalloproteinase (MMP)-9 activity. This study suggests that activin A signaling may contribute to the pathology of bronchopulmonary dysplasia.
Sigma-Aldrich
Follistatin human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
6-Maleimidohexanoic acid, 90% (GC)