Merck
  • Rhinoviral stimuli, epithelial factors and ATP signalling contribute to bronchial smooth muscle production of IL-33.
Rhinoviral stimuli, epithelial factors and ATP signalling contribute to bronchial smooth muscle production of IL-33.
Journal of translational medicine (2015-09-01)
Jenny Calvén, Hamid Akbarshahi, Mandy Menzel, Cemil Korcan Ayata, Marco Idzko, Leif Bjermer, Lena Uller
ABSTRACT
Bronchial smooth muscle cells (BSMCs) from severe asthmatics have been shown to overexpress the Th2-driving and asthma-associated cytokine IL-33. However, little is known regarding factors involved in BSMC production of IL-33. Rhinovirus (RV) infections cause asthma exacerbations, which exhibit features of Th2-type inflammation. Here, we investigated the effects of epithelial-derived media and viral stimuli on IL-33 expression in human BSMCs. Primary human BSMCs from healthy (n = 3) and asthmatic (n = 3) subjects were stimulated with conditioned media from primary human bronchial epithelial cells (BECs), double-stranded (ds)RNA, dsRNA/LyoVec, or infected with RV. BSMCs were also pretreated with the purinergic receptor antagonist suramin. IL-33 expression was analysed by RT-qPCR and western blot and ATP levels were determined in cell supernatants. RV infection and activation of TLR3 by dsRNA increased IL-33 mRNA and protein in healthy and asthmatic BSMCs. These effects were inhibited by dexamethasone. BSMC expression of IL-33 was also increased by stimulation of RIG-I-like receptors using dsRNA/LyoVec. Conditioned media from BECs induced BSMC expression of IL-33, which was further enhanced by dsRNA. BEC-derived medium and viral-stimulated BSMC supernatants exhibited elevated ATP levels. Blocking of purinergic signalling with suramin inhibited BSMC expression of IL-33 induced by dsRNA and BEC-derived medium. RV infection of BSMCs and activation of TLR3 and RIG-I-like receptors cause expression and production of IL-33. Epithelial-released factor(s) increase BSMC expression of IL-33 and exhibit positive interaction with dsRNA. Increased BSMC IL-33 associates with ATP release and is antagonised by suramin. We suggest that epithelial-derived factors contribute to baseline BSMC IL-33 production, which is further augmented by RV infection of BSMCs and stimulation of their pathogen-recognising receptors.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder