ชื่อสินค้า
Diisopropylamine puriss. p.a., ≥99.0% (GC)
หมายเลขสินค้า
38290
ยี่ห้อสินค้า
SIAL
CASE番号
108-18-9
น้ำหนักโมเลกุล
101.19
ทดสอบ | ข้อกำหนด |
|---|---|
| APPEARANCE (COLOR) | Colorless |
| APPEARANCE (FORM) | Liquid |
| PURITY (GC AREA %) | ≥ 99.0 % |
| REFRACTIVE INDEX N20/D | 1.392 - 1.394 |
| WATER | ≤ 0.5 % |
| RESIDUE (EVAPORATION) | ≤ 0.001 % |
| PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
| METAL TRACE ANALYSIS (ICP) | Complying |
| ALUMINIUM (ICP) | ≤ 0.5 ppm |
| BARIUM (ICP) | ≤ 0.1 ppm |
| BISMUTH (ICP) | ≤ 0.1 ppm |
| CALCIUM (ICP) | ≤ 0.5 ppm |
| CADMIUM (ICP) | ≤ 0.05 ppm |
| COBALT (ICP) | ≤ 0.02 ppm |
| CHROMIUM (ICP) | ≤ 0.02 ppm |
| COPPER (ICP) | ≤ 0.02 ppm |
| IRON (ICP) | ≤ 0.5 ppm |
| POTASSIUM (ICP) | ≤ 0.5 ppm |
| LITHIUM (ICP) | ≤ 0.1 ppm |
| MAGNESIUM (ICP) | ≤ 0.1 ppm |
| MANGANESE (ICP) | ≤ 0.02 ppm |
| MOLYBDENUM (ICP) | ≤ 0.1 ppm |
| SODIUM (ICP) | ≤ 0.5 ppm |
| NICKEL (ICP) | ≤ 0.02 ppm |
| LEAD (ICP) | ≤ 0.1 ppm |
| STRONTIUM (ICP) | ≤ 0.1 ppm |
| ZINC (ICP) | ≤ 0.1 ppm |