Merck
  • Evidence that the Gh protein is a signal mediator from alpha 1-adrenoceptor to a phospholipase C. II. Purification and characterization of a Gh-coupled 69-kDa phospholipase C and reconstitution of alpha 1-adrenoceptor, Gh family, and phospholipase C.
Evidence that the Gh protein is a signal mediator from alpha 1-adrenoceptor to a phospholipase C. II. Purification and characterization of a Gh-coupled 69-kDa phospholipase C and reconstitution of alpha 1-adrenoceptor, Gh family, and phospholipase C.
The Journal of biological chemistry (1993-12-25)
T Das, K J Baek, C Gray, M J Im
ABSTRACT
Our studies on the alpha 1-adrenoceptor signaling have demonstrated that the Gh family is a signal mediator. We report here that a 69-kDa phospholipase C (PLC) is the effector in this signal pathway. The enzyme was isolated by dissociating a Gh7-PLC complex which was induced in the bovine liver membranes incubating with (-)-epinephrine and GTP. The enzyme displayed a marked preference hydrolysis for phosphatidylinositol 4,5-bisphosphate over other phosphatidylinositides at micromolar calcium. Reconstitution of PLC with the alpha 1-adrenoceptor and Gh (Gh7) into phospholipid vesicles resulted in a lowered Ca2+ requirement for the substrate hydrolysis in the presence of guanosine 5'-3-O-(thio)triphosphate (GTP gamma S) when the receptor was activated with the alpha 1-agonist. The formation of inositol phosphate was hormone concentration dependent and reached maximal within 3 min which was faster than the formation in the presence of the alpha 1-antagonist. An Gh7 alpha antibody co-immunoprecipitated 80-85% of phospholipase C activity in the presence of GTP gamma S, but not in the presence of GDP or buffer, showing the association of PLC with the alpha-subunit of Gh family. Thus, our novel approaches to identify the effector involved in the alpha 1-adrenoceptor signaling, as well as the reconstitution studies, substantially demonstrate that the alpha 1-adrenoceptor-mediated transmembrane signaling involves the Gh family and a 69-kDa PLC.
Sigma-Aldrich
Phospholipase C from Bacillus cereus, ≥200 units/mg protein
Pricing and availability is not currently available.