Merck
  • Biomarker discovery in non-small cell lung cancer: integrating gene expression profiling, meta-analysis, and tissue microarray validation.
Biomarker discovery in non-small cell lung cancer: integrating gene expression profiling, meta-analysis, and tissue microarray validation.
Clinical cancer research : an official journal of the American Association for Cancer Research (2012-10-04)
Johan Botling, Karolina Edlund, Miriam Lohr, Birte Hellwig, Lars Holmberg, Mats Lambe, Anders Berglund, Simon Ekman, Michael Bergqvist, Fredrik Pontén, André König, Oswaldo Fernandes, Mats Karlsson, Gisela Helenius, Christina Karlsson, Jörg Rahnenführer, Jan G Hengstler, Patrick Micke
ABSTRACT
Global gene expression profiling has been widely used in lung cancer research to identify clinically relevant molecular subtypes as well as to predict prognosis and therapy response. So far, the value of these multigene signatures in clinical practice is unclear, and the biologic importance of individual genes is difficult to assess, as the published signatures virtually do not overlap. Here, we describe a novel single institute cohort, including 196 non-small lung cancers (NSCLC) with clinical information and long-term follow-up. Gene expression array data were used as a training set to screen for single genes with prognostic impact. The top 450 probe sets identified using a univariate Cox regression model (significance level P < 0.01) were tested in a meta-analysis including five publicly available independent lung cancer cohorts (n = 860). The meta-analysis revealed 14 genes that were significantly associated with survival (P < 0.001) with a false discovery rate <1%. The prognostic impact of one of these genes, the cell adhesion molecule 1 (CADM1), was confirmed by use of immunohistochemistry on tissue microarrays from 2 independent NSCLC cohorts, altogether including 617 NSCLC samples. Low CADM1 protein expression was significantly associated with shorter survival, with particular influence in the adenocarcinoma patient subgroup. Using a novel NSCLC cohort together with a meta-analysis validation approach, we have identified a set of single genes with independent prognostic impact. One of these genes, CADM1, was further established as an immunohistochemical marker with a potential application in clinical diagnostics.
Sigma-Aldrich
Anti-SynCAM antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.