Merck
GC Analysis of QA Test Mix on SLB®-IL59 using Air as Carrier Gas, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of QA Test Mix on SLB®-IL59 using Air as Carrier Gas, Comparison to a Polyethylene Glycol (PEG) Column suitable for GC
Materials
related product
Product No.
Description
Pricing
SLB®-IL59 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL59, 30 m x 0.25 mm I.D., 0.20 μm (28891-U)
column
PEG phase, 30 m x 0.25 mm I.D., 0.25 μm
oven
50 °C (2 min), 4 °C/min to 200 °C (15 min)
inj. temp.
250 °C
detector
250 °C
carrier gas
compressed air, 16 psi constant
injection
1 μL 50:1 split
sample
Programmed Test Mix
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany