Merck
HPLC Analysis of Proteins by Size Exclusion on Zenix® and SRT® SEC-300, Comparison of Particle Size and Column I.D.
HPLC Analysis of Proteins by Size Exclusion on Zenix® and SRT® SEC-300, Comparison of Particle Size and Column I.D. application for HPLC
Materials
related product
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 4.6 mm, 3 μm particle size, stainless steel
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
SRT® SEC-300 Gel Filtration Column, 5 μm
L × I.D. 30 cm × 7.8 mm, 5 μm particle size, stainless steel
CONDITIONS
column
Zenix 300, 30 cm x 4.6 mm I.D., 3 μm particles, 300 Å (Z777028)
column
Zenix 300, 30 cm x 7.8 mm I. D., 3 μm particles, 300 Å (Z777033)
column
SRT SEC-300, 30 cm x 7.8 mm 5 μm particles, 300 Å (Z777051)
mobile phase
150 mM sodium phosphate, pH 7
column temp.
ambient
detector
UV, 214nm
sample
each compound 1 mg/mL except uracil, 2.5 μg/mL
Description
Legal Information
SRT is a registered trademark of Sepax Technologies
Zenix is a registered trademark of Sepax Technologies