Merck
LC/MS/MS Analysis of Zopiclone and Metabolites on Ascentis® Express Phenyl-Hexyl
LC/MS/MS Analysis of Zopiclone and Metabolites on Ascentis® Express Phenyl-Hexyl application for LC-MS
Materials
related product
Product No.
Description
Pricing
Ascentis® Express Phenyl-Hexyl (2.7 μm) HPLC Column
L × I.D. 10 cm × 2.1 mm, pk of 1 ea
Zopiclone solution
1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Zopiclone-N-oxide solution
100 μg/mL (Methanol with 1% 1 M HCl), ampule of 1 mL, certified reference material, Cerilliant®
N-Desmethylzopiclone solution
100 μg/mL in acetonitrile, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express Phenyl-Hexyl  10 cm x 2.1 mm, 2.7 μm particles (53336-U)
mobile phase
[A] 13 mM ammonium acetate in water, no pH adjustment; [B] 13 mM ammonium acetate in methanol; (52:48, A:B)
flow rate
0.4 mL/min
pressure
7850 psi (541 bar)
column temp.
35 °C
detector
MS, ESI(+), MRM
injection
1 μL
sample
250 ng/mL each in 13 mM ammonium acetate in water, no pH adjustment; [B] 13 mM ammonium acetate in methanol; (50:50 A:B)
Description
Analysis Note
Zopiclone is a non-benzodiazepine hypnotic sold under the trade names Imovane® and Zimovane and is prescribed for the treatment of insomnia. The Ascentis Express phenyl-Hexyl column gave the best separation of zopiclone and its metabolites of the column chemistries tested. Ultra high purity LC-MS solvents were used to supply low background interference and low particulate contaminants for robust, trouble-free operation. Cerilliant CRMs provided reliable identification and quantification.
Other Notes
MRM Transitions
1  : 375.11 > 111.92      N-Desmethylzopiclone
2  : 375.11 > 217.00       N-Desmethylzopiclone
3  : 375.11 > 244.98       N-Desmethylzopiclone
4  : 389.08 > 111.92       Zopiclone           
5  : 389.08 > 217.00       Zopiclone           
6  : 389.08 > 244.97       Zopiclone         
7  : 405.13 > 58.01         Zopiclone-N-oxide   
8  : 405.13 > 143.29       Zopiclone-N-oxide   
9  : 405.13 > 245.03      Zopiclone-N-oxide
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Imovane is a registered trademark of Sanofi-Aventis