Skip to Content
MilliporeSigma

BDPH9BCC32CB

Bide Pharmatech

(4-Bromo-2-ethylphenyl)methanol

Synonym(s):

(4-bromo-2-ethylphenyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1Br1
CAS Number:
Molecular Weight:
215.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11BrO/c1-2-7-5-9(10)4-3-8(7)6-11/h3-5,11H,2,6H2,1H3

Inchi Key

JCUFCMGXQKHDLU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?

Explore all of our products under (4-Bromo-2-ethylphenyl)methanol



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library