Skip to Content
MilliporeSigma

BDPH9BCC68DB

Bide Pharmatech

3-(Benzo[d]thiazol-2-yl)aniline

Synonym(s):

3-(1,3-benzothiazol-2-yl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H10N2S1
Molecular Weight:
226.3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H10N2S/c14-10-5-3-4-9(8-10)13-15-11-6-1-2-7-12(11)16-13/h1-8H,14H2

Inchi Key

MKMRZZJJDIBAJF-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library