Skip to Content
MilliporeSigma

BDPH9BCC720F

Bide Pharmatech

7-Bromo-1,5-naphthyridin-3-amine

Synonym(s):

7-bromo-1,5-naphthyridin-3-amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6N3Br1
CAS Number:
Molecular Weight:
224.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6BrN3/c9-5-1-7-8(11-3-5)2-6(10)4-12-7/h1-4H,10H2

Inchi Key

UUUFOHPAGLDYLC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to yellow-brown solid



Still not finding the right product?

Explore all of our products under 7-Bromo-1,5-naphthyridin-3-amine


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library